MilliporeSigma
  • Are mast cells still good biomarkers for bladder pain syndrome/interstitial cystitis?
Are mast cells still good biomarkers for bladder pain syndrome/interstitial cystitis?
The Journal of urology (2015-01-18)
Marianne Gamper, Sigrid Regauer, JoEllen Welter, Jakob Eberhard, Volker Viereck
ABSTRACT
ESSIC identifies mast cell infiltrates of detrusor muscle as a diagnostic criterion for bladder pain syndrome/interstitial cystitis. However, an increased mast cell count is also characteristic of overactive bladder syndrome. The lack of uniformity in mast cell detection methods hampers data comparison. Using state-of-the-art techniques we investigated whether mast cells differ among bladder conditions. We analyzed bladder biopsies from 56 patients, including 31 with bladder pain syndrome/interstitial cystitis with (12) or without (19) Hunner lesions, 13 with overactive bladder syndrome and 12 without bladder symptoms to determine the quantity, location, distribution and activation of mast cells using immunohistochemistry with anti-mast cell tryptase. Patients were allocated to study groups by key bladder symptoms commonly used to define conditions (pain and major urgency). Subepithelial mast cell localization (p <0.001) and an increased detrusor mast cell count (p = 0.029) were characteristic of bladder pain syndrome/interstitial cystitis with Hunner lesions. The optimal cutoff of 32 detrusor mast cells per mm(2) achieved only 68% accuracy with 38% positive predictive value. No difference was observed between bladder pain syndrome/interstitial cystitis without Hunner lesions and overactive bladder syndrome. Patient groups differed in lymphocyte infiltration (p = 0.001), nodular lymphocyte aggregates (p <0.001) and urothelium integrity (p <0.001). Subepithelial mast cell distribution was characteristic of bladder pain syndrome/interstitial cystitis with Hunner lesions. Detrusor mastocytosis had poor predictive value for bladder pain syndrome/interstitial cystitis. Mast cell assessment did not distinguish bladder pain syndrome/interstitial cystitis without Hunner lesions from overactive bladder syndrome.
Sigma-Aldrich
MISSION® esiRNA, targeting human TPSD1