MilliporeSigma
  • Effects of nitric oxide synthase deficiency on a disintegrin and metalloproteinase domain-containing protein 12 expression in mouse brain samples.
Effects of nitric oxide synthase deficiency on a disintegrin and metalloproteinase domain-containing protein 12 expression in mouse brain samples.
Molecular medicine reports (2015-04-22)
Uwe Lendeckel, Carmen Wolke, Hans-Gert Bernstein, Gerburg Keilhoff
ABSTRACT
A disintegrin and metalloproteinase domain-containing protein 12 (ADAM12) belongs to the ADAM family of transmembrane proteins. Via proteolysis, cell adhesion, cell-cell fusion, cell-matrix interaction and membrane protein shedding, ADAM proteins are involved in normal brain development, and also in cancer genesis and progression, and in inflammation. Therefore, neurobiological research focusing on this protein is increasing. Nitric oxide (NO), which is endogenously produced by NO synthases (NOS), is associated with glial tumors. However, knock-out of NOS produces only limited antitumor effects. The present study analyzed the expression of ADAM12 in the cortex and hippocampus of C57/BL6 wild-type mice, and endothelial NOS-, neuronal NOS-(nNOS) or inducible NOS (iNOS)-deficient (-/-) mice, at different stages of development. Expression of ADAM12 was quantified using immunoblot analysis of cortical and hippocampal tissue samples from fetal, neonatal (5 days postnatal), adult (12 weeks old) or >1 year old mice. Using reverse transcription-quantitative polymerase chain reaction, ADAM12 expression was analyzed in cultured N9, OLN93, C6 and PC12 cells, representing the four main cell types in the brain, following NOS inhibition. ADAM12 expression was low in all mouse genotypes and regions of the brain, and in fetal and neonatal mice, an increase in expression was observed with increasing age. The highest levels of expression were observed in the cortex of adult mice, iNOS(-/-) mice of >1 year and wild-type mice, and in the hippocampus of adult and iNOS(-/-) mice of >1 year. By contrast, ADAM12 expression was lowest in adult nNOS(-/-) mice. Inhibition of NOS using N(ω)-Nitro-L-arginine methyl ester hydrochloride, induced ADAM12 mRNA expression in N9 and PC12 cell lines. Inhibition of NOS using L-N(6)-(1-Iminoethyl)lysine dihydrochloride, induced ADAM12 mRNA expression in N9 and C6 cell lines. No change in ADAM12 expression was observed in OLN93 cells following NOS inhibition. ADAM12 expression in mouse hippocampus and cortex samples demonstrated considerable variation during development, with a marked increase observed in adult and >1 year old mice, compared with that in fetal and neonatal mice.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Chloromethane solution, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Chloromethane solution, 1.0 M in diethyl ether
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
L-N6-(1-Iminoethyl)lysine dihydrochloride
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%