MilliporeSigma
  • MircoRNA-33a inhibits epithelial-to-mesenchymal transition and metastasis and could be a prognostic marker in non-small cell lung cancer.
MircoRNA-33a inhibits epithelial-to-mesenchymal transition and metastasis and could be a prognostic marker in non-small cell lung cancer.
Scientific reports (2015-09-04)
Lihua Yang, Jie Yang, Jingqiu Li, Xingkai Shen, Yanping Le, Chengwei Zhou, Shaomin Wang, Shun Zhang, Dazhi Xu, Zhaohui Gong
ABSTRACT
Understanding the molecular mechanism by which epithelial mesenchymal transition (EMT)-mediated cancer metastasis and how microRNA (miRNA) regulates lung cancer progression via Twist1-activated EMT may provide potential therapeutic targets for cancer therapy. Here we found that miR-33a, an intronic miRNA located within the sterol regulatory element-binding protein 2 (SREBP-2) gene, is expressed at low levels in metastatic non-small cell lung cancer (NSCLC) cells and is inversely correlated with Twist1 expression. Conversely, miR-33a knockdown induces EMT and miR-33a overexpression blocks EMT by regulating of Twist1 expression in NSCLC cells. Bioinformatical prediction and luciferase reporter assay confirm that Twist1 is a direct target of miR-33a. Additionally, Twist1 knockdown blocks EMT-related metastasis and forced expression of miR-33a inhibits lung cancer metastasis in a xenograft animal model. Clinically, miR-33a is found to be at low levels in NSCLC patients and down-regulation of miR-33a predicts a poor prognosis. These findings suggest that miR-33a targets Twist1 and inhibits invasion and metastasis in NSCLC. Thus, miR-33a might be a potential prognostic marker and of therapeutic relevance for NSCLC metastasis intervention.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Twist1
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human TWIST1
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%