MilliporeSigma
  • Axin Regulates Dendritic Spine Morphogenesis through Cdc42-Dependent Signaling.
Axin Regulates Dendritic Spine Morphogenesis through Cdc42-Dependent Signaling.
PloS one (2015-07-24)
Yu Chen, Zhuoyi Liang, Erkang Fei, Yuewen Chen, Xiaopu Zhou, Weiqun Fang, Wing-Yu Fu, Amy K Y Fu, Nancy Y Ip
ABSTRACT
During development, scaffold proteins serve as important platforms for orchestrating signaling complexes to transduce extracellular stimuli into intracellular responses that regulate dendritic spine morphology and function. Axin ("axis inhibitor") is a key scaffold protein in canonical Wnt signaling that interacts with specific synaptic proteins. However, the cellular functions of these protein-protein interactions in dendritic spine morphology and synaptic regulation are unclear. Here, we report that Axin protein is enriched in synaptic fractions, colocalizes with the postsynaptic marker PSD-95 in cultured hippocampal neurons, and interacts with a signaling protein Ca(2+)/calmodulin-dependent protein kinase II (CaMKII) in synaptosomal fractions. Axin depletion by shRNA in cultured neurons or intact hippocampal CA1 regions significantly reduced dendritic spine density. Intriguingly, the defective dendritic spine morphogenesis in Axin-knockdown neurons could be restored by overexpression of the small Rho-GTPase Cdc42, whose activity is regulated by CaMKII. Moreover, pharmacological stabilization of Axin resulted in increased dendritic spine number and spontaneous neurotransmission, while Axin stabilization in hippocampal neurons reduced the elimination of dendritic spines. Taken together, our findings suggest that Axin promotes dendritic spine stabilization through Cdc42-dependent cytoskeletal reorganization.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Axin1
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human AXIN1
Pricing and availability is not currently available.