MilliporeSigma
  • Artesunate induces apoptosis via a ROS-independent and Bax-mediated intrinsic pathway in HepG2 cells.
Artesunate induces apoptosis via a ROS-independent and Bax-mediated intrinsic pathway in HepG2 cells.
Experimental cell research (2015-07-15)
Guiqi Qin, Liping Wu, Hongyu Liu, Yilin Pang, Chubiao Zhao, Shengnan Wu, Xiaoping Wang, Tongsheng Chen
ABSTRACT
This study aims to explore the detail molecular mechanism by which artesunate (ARS), an artemisinin derivative, induces apoptosis in HepG2 cells. ARS induced a loss of mitochondrial transmemberane potential (ΔΨm), phosphatidylserine (PS) externalization, as well as activations of Bax/Bak and caspases indicative of apoptosis induction. Silencing Bax but not Bak significantly inhibited ARS-induced apoptosis, demonstrating the key role of the Bax-mediated intrinsic pathway. Although ARS increased intracellular reactive oxygen species (ROS), ARS-induced apoptosis was neither prevented by pretreatment with ROS scavengers nor potentiated by pretreatment with l-buthionine-sulfoximine (BSO) that enhanced the ARS-induced intracellular ROS generation, demonstrating that ROS was not involved in ARS-induced apoptosis. In addition, ARS did not induce Bid translocation to mitochondria, and the cytotoxicity of ARS was not prevented by silencing Bim, Puma or Mcl-1, but was significantly enhanced by HA14-1 pretreatment, demonstrating that Bcl-2/-xl instead of Bid and Bim as well as Puma may be the upstream factor to regulate the Bax-mediated intrinsic pathway. Collectively, our data demonstrate that ARS induces ROS-independent apoptosis via the Bax-mediated intrinsic pathway in HepG2 cells.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%