MilliporeSigma
  • Protection by sigma-1 receptor agonists is synergic with donepezil, but not with memantine, in a mouse model of amyloid-induced memory impairments.
Protection by sigma-1 receptor agonists is synergic with donepezil, but not with memantine, in a mouse model of amyloid-induced memory impairments.
Behavioural brain research (2015-09-20)
Tangui Maurice
ABSTRACT
Drugs activating the sigma-1 (σ1) chaperone protein are anti-amnesic and neuroprotective in neurodegenerative pathologies like Alzheimer's disease (AD). Since these so-called σ1 receptor (σ1R) agonists modulate cholinergic and glutamatergic systems in a variety of physiological responses, we addressed their putative additive/synergistic action in combination with cholinergic or glutamatergic drugs. The selective σ1 agonist PRE-084, or the non-selective σ1 drug ANAVEX2-73 was combined with the acetylcholinesterase inhibitor donepezil or the NMDA receptor antagonist memantine in the nontransgenic mouse model of AD-like memory impairments induced by intracerebroventricular injection of oligomeric Aβ25-35 peptide. Two behavioral tests, spontaneous alternation and passive avoidance response, were used in parallel and both protective and symptomatic effects were examined. After determination of the minimally active doses for each compound, the combinations were tested and the combination index (CI) calculated. Combinations between the σ1 agonists and donepezil showed a synergic protective effect, with CI<1, whereas the combinations with memantine showed an antagonist effect, with CI>1. Symptomatic effects appeared only additive for all combinations, with CI=1. A pharmacological analysis of the PRE-084+donepezil combination revealed that the synergy could be due to an inter-related mechanism involving α7 nicotinic ACh receptors and σ1R. These results demonstrated that σ1 drugs do not only offer a protective potential alone but also in combination with other therapeutic agents. The nature of neuromodulatory molecular chaperone of the σ1R could eventually lead to synergistic combinations.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
PRE-084, solid
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid