MilliporeSigma
  • Gas-Phase Fragmentation Pathways of Mixed Addenda Keggin Anions: PMo12-nW nO 40 3- (n = 0-12).
Gas-Phase Fragmentation Pathways of Mixed Addenda Keggin Anions: PMo12-nW nO 40 3- (n = 0-12).
Journal of the American Society for Mass Spectrometry (2015-04-03)
K Don D Gunaratne, Venkateshkumar Prabhakaran, Grant E Johnson, Julia Laskin
ABSTRACT
We report a collision-induced dissociation (CID) investigation of the mixed addenda polyoxometalate (POM) anions, PMo(12-n)W(n)O(40)(3-) (n = 0-12). The anions were generated in solution using a straightforward single-step synthesis approach and introduced into the gas phase by electrospray ionization (ESI). Distinct differences in fragmentation patterns were observed for the range of mixed addenda POMs examined in this study. CID of molybdenum-rich anions, PMo(12-n)W(n)O(40)(3-) (n = 0-2), generates an abundant doubly charged fragment containing seven metal atoms (M) and 22 oxygen atoms (M(7)O(22)(2-)) and its complementary singly charged PM(5)O(18)(-) ion. In comparison, the doubly charged Lindqvist anion, (M(6)O(19)(2-)) and its complementary singly charged PM(6)O(21)(-) ion are the dominant fragments of Keggin POMs containing more than two tungsten atoms, PMo(12-n)W(n)O(40)(3-) (n = 3-12). The observed transition in the dissociation pathways with an increase in the number of W atoms in the POM may be attributed to the higher barrier of tungsten-rich anions towards isomerization. We present evidence that the observed distribution of Mo and W atoms in the major M(6)O(19)(2-) and M(7)O(22)(2-) fragment ions is different from that predicted by a random distribution, indicating substantial segregation of the addenda metal atoms in the POMs. Charge reduction of the triply charged precursor anion resulting in formation of doubly charged anions is also observed. This is a dominant pathway for mixed POMs having a majority (8-11) of W atoms and a minor channel for other precursors indicating a close competition between fragmentation and charge loss pathways in CID of POM anions.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphomolybdate hydrate, technical
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium phosphotungstate hydrate, ≥99.9% trace metals basis