Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Pricing and availability is not currently available.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.