MilliporeSigma
  • Nuclear factor erythroid 2-related factor 2 (Nrf2) regulates airway epithelial barrier integrity.
Nuclear factor erythroid 2-related factor 2 (Nrf2) regulates airway epithelial barrier integrity.
Allergology international : official journal of the Japanese Society of Allergology (2015-09-08)
Yoshitaka Shintani, Shuichiro Maruoka, Yasuhiro Gon, Daisuke Koyama, Akiko Yoshida, Yutaka Kozu, Kazumichi Kuroda, Ikuko Takeshita, Eriko Tsuboi, Kaori Soda, Shu Hashimoto
ABSTRACT
Inhaled corticosteroids enhance airway epithelial barrier integrity. However, the mechanism by which they accomplish this is unclear. Therefore, we investigated steroid-inducible genes and signaling pathways that were involved in enhancing airway epithelial barrier integrity. A human bronchial epithelial cell line (16HBE cells) was cultured with 10(-6) M dexamethasone (DEX) for 3 days to enhance epithelial barrier integrity. After measuring transepithelial electrical resistance (TER) and paracellular permeability, we extracted total RNA from 16HBE cells and performed microarray and pathway analysis. After we identified candidate genes and a canonical pathway, we measured TER and immunostained for tight junction (TJ) and adherent junction (AJ) proteins in cells that had been transfected with specific small interfering RNAs (siRNAs) for these genes. We identified a nuclear factor erythroid 2-related factor 2 (Nrf2)-mediated oxidative stress response pathway which was primarily involved in the steroid-induced enhancement of airway epithelial barrier integrity. Transfecting cells with Nrf2 specific siRNA reduced the steroid-induced enhancement of airway epithelial barrier integrity and the accumulation of TJ and AJ proteins at sites of cell-cell contact. Moreover, based on pathway analysis, aldehyde oxidase 1 (AOX1) was identified as a downstream enzyme of Nrf2. Transfecting cells with AOX1-specific siRNA also reduced the steroid-induced enhancement of airway epithelial barrier integrity. Our results indicated that the Nrf2/AOX1 pathway was important for enhancing airway epithelial barrier integrity. Because the airway epithelium of asthmatics is susceptible to reduced barrier integrity, this pathway might be a new therapeutic target for asthma.
Sigma-Aldrich
MISSION® esiRNA, targeting human T
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)