MilliporeSigma
  • Placental Fatty Acid ethyl esters are elevated with maternal alcohol use in pregnancies complicated by prematurity.
Placental Fatty Acid ethyl esters are elevated with maternal alcohol use in pregnancies complicated by prematurity.
PloS one (2015-05-16)
Theresa W Gauthier, Sowmya S Mohan, Teresa S Gross, Frank L Harris, David M Guidot, Lou Ann S Brown
ABSTRACT
The accumulation of fatty acid ethyl esters (FAEEs) in meconium of term newborns has been described as one potential biomarker of maternal alcohol use during pregnancy. FAEEs accumulate in multiple alcohol-exposed fetal tissues and in the placenta. Limited research has focused on the identification of the premature newborn exposed to alcohol in utero. We hypothesized that maternal alcohol use occurs in a significant proportion of premature deliveries and that this exposure can be detected as elevated placental FAEEs. The goals of this study were to 1) determine the prevalence of maternal alcohol use in the premature newborn and 2) investigate whether placental FAEEs could identify those newborns with fetal alcohol exposure. This prospective observational study evaluated 80 placentas from 80 women after premature delivery. Subjects were interviewed for alcohol intake and placental FAEEs were quantified via GC/MS. Receiver Operator Characteristic (ROC) Curves were generated to evaluate the ability of placental FAEEs to predict maternal drinking during pregnancy. Adjusted ROC curves were generated to adjust for gestational age, maternal smoking, and illicit drug use. 30% of the subjects admitted to drinking alcohol during pregnancy and approximately 14% answered questions indicative of problem drinking (designated AUDIT+). The specific FAEEs ethyl stearate and linoleate, as well as combinations of oleate + linoleate + linolenate (OLL) and of OLL + stearate, were significantly (p<0.05) elevated in placentas from AUDIT+ pregnancies. Adjusted ROC Curves generated areas under the curve ranging from 88-93% with negative predictive values of 97% for AUDIT+ pregnancies. We conclude that nearly one third of premature pregnancies were alcohol-exposed, and that elevated placental FAEEs hold great promise to accurately determine maternal alcohol use, particularly heavy use, in pregnancies complicated by premature delivery.
Sigma-Aldrich
Ethyl heptadecanoate, AldrichCPR