MilliporeSigma
  • A new sensitive determination method of estradiol in plasma using peroxyoxalate ester chemiluminescence combined with an HPLC system.
A new sensitive determination method of estradiol in plasma using peroxyoxalate ester chemiluminescence combined with an HPLC system.
Biomedical chromatography : BMC (2000-08-29)
H Yamada, Y Kuwahara, Y Takamatsu, T Hayase
ABSTRACT
A new sensitive determination method of estradiol in a plasma sample using peroxyoxalate ester chemiluminescence was developed. Estradiol, which was extracted by liquid-liquid extraction using ethyl acetate from plasma, was derivatized with dansyl-chloride (DNS-Cl) and separated by reverse-phase HPLC. The performance of four oxalates, bis(trichlorophenyl)oxalate (TCPO), bis(2,4-dinitrophenyl)oxalate (DNPO), bis(pentafluorophenyl)oxalate (PFPO), and bis[4-nitro-2-(3,6, 9-trioxadecyloxycarbonyl)phenyl] oxalate (TDPO), were evaluated using the static system, and DNPO was found to have the most sensitive and stable chemiluminescence at a H(2)O(2) concentration of 30 mM. HPLC-chemiluminescence system using DNPO for the determination of estradiol was established. The detection limit of dansylated-estradiol (DNS-E2) was 15 fmol (4 pg) in the standard solution and 44 fmol (12 pg) in the rat plasma sample at S/N = 3.