MilliporeSigma
  • A randomized, controlled phase III trial of nab-Paclitaxel versus dacarbazine in chemotherapy-naïve patients with metastatic melanoma.
A randomized, controlled phase III trial of nab-Paclitaxel versus dacarbazine in chemotherapy-naïve patients with metastatic melanoma.
Annals of oncology : official journal of the European Society for Medical Oncology (2015-09-28)
E M Hersh, M Del Vecchio, M P Brown, R Kefford, C Loquai, A Testori, S Bhatia, R Gutzmer, R Conry, A Haydon, C Robert, S Ernst, J Homsi, J J Grob, K Kendra, S S Agarwala, M Li, A Clawson, C Brachmann, M Karnoub, I Elias, M F Renschler, A Hauschild
ABSTRACT
The efficacy and safety of nab-paclitaxel versus dacarbazine in patients with metastatic melanoma was evaluated in a phase III randomized, controlled trial. Chemotherapy-naïve patients with stage IV melanoma received nab-paclitaxel 150 mg/m(2) on days 1, 8, and 15 every 4 weeks or dacarbazine 1000 mg/m(2) every 3 weeks. The primary end point was progression-free survival (PFS) by independent radiologic review; the secondary end point was overall survival (OS). A total of 529 patients were randomized to nab-paclitaxel (n = 264) or dacarbazine (n = 265). Baseline characteristics were well balanced. The majority of patients were men (66%), had an Eastern Cooperative Oncology Group status of 0 (71%), and had M1c stage disease (65%). The median PFS (primary end point) was 4.8 months with nab-paclitaxel and 2.5 months with dacarbazine [hazard ratio (HR), 0.792; 95.1% confidence interval (CI) 0.631-0.992; P = 0.044]. The median OS was 12.6 months with nab-paclitaxel and 10.5 months with dacarbazine (HR, 0.897; 95.1% CI 0.738-1.089; P = 0.271). Independently assessed overall response rate was 15% versus 11% (P = 0.239), and disease control rate (DCR) was 39% versus 27% (P = 0.004) for nab-paclitaxel versus dacarbazine, respectively. The most common grade ≥3 treatment-related adverse events were neuropathy (nab-paclitaxel, 25% versus dacarbazine, 0%; P < 0.001), and neutropenia (nab-paclitaxel, 20% versus dacarbazine, 10%; P = 0.004). There was no correlation between secreted protein acidic and rich in cysteine (SPARC) status and PFS in either treatment arm. nab-Paclitaxel significantly improved PFS and DCR compared with dacarbazine, with a manageable safety profile.
Sigma-Aldrich
Dacarbazine, antineoplastic purine analog
Pricing and availability is not currently available.