MilliporeSigma
  • Dual Role of the 1,2,3-Triazolium Ring as a Hydrogen-Bond Donor and Anion-π Receptor in Anion-Recognition Processes.
Dual Role of the 1,2,3-Triazolium Ring as a Hydrogen-Bond Donor and Anion-π Receptor in Anion-Recognition Processes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-03)
Fabiola Zapata, Lidia Gonzalez, Antonio Caballero, Ibón Alkorta, Jose Elguero, Pedro Molina
ABSTRACT
Several bis(triazolium)-based receptors have been synthesized as chemosensors for anion recognition. The central naphthalene core features two aryltriazolium side-arms. NMR experiments revealed differences between the binding modes of the two triazolium rings: one triazolium ring acts as a hydrogen-bond donor, the other as an anion-π receptor. Receptors 9(2+)⋅2BF4(-) (C6H5), 11(2+)⋅2BF4(-) (4-NO2-C6H4), and 13(2+)⋅2BF(4-) (ferrocenyl) bind HP2O7(3-) anions in a mixed-binding mode that features a combination of hydrogen-bonding and anion-π interactions and results in strong binding. On the other hand, receptor 10(2+)⋅2 BF4(-) (4-CH3O-C6H4) only displays combined Csp2-H/anion-π interactions between the two arms of the receptors and the bound anion rather than triazolium (CH)(+)⋅⋅⋅anion hydrogen bonding. All receptors undergo a downfield shift of the triazolium protons, as well as the inner naphthalene protons, in the presence of H2PO4(-) anions. That suggests that only hydrogen-bonding interactions exist between the binding site and the bound anion, and involve a combination of cationic (triazolium) and neutral (naphthalene) C-H donor interactions. Theoretical calculations relate the electronic structure of the substituent on the aromatic group with the interaction energies and provide a minimum-energy conformation for all the complexes that explains their measured properties.
Sigma-Aldrich
1-Ethynyl-4-nitrobenzene, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%
Pricing and availability is not currently available.