MilliporeSigma
  • Supramolecular regulation of bioorthogonal catalysis in cells using nanoparticle-embedded transition metal catalysts.
Supramolecular regulation of bioorthogonal catalysis in cells using nanoparticle-embedded transition metal catalysts.
Nature chemistry (2015-06-24)
Gulen Yesilbag Tonga, Youngdo Jeong, Bradley Duncan, Tsukasa Mizuhara, Rubul Mout, Riddha Das, Sung Tae Kim, Yi-Cheun Yeh, Bo Yan, Singyuk Hou, Vincent M Rotello
ABSTRACT
Bioorthogonal catalysis broadens the functional possibilities of intracellular chemistry. Effective delivery and regulation of synthetic catalytic systems in cells are challenging due to the complex intracellular environment and catalyst instability. Here, we report the fabrication of protein-sized bioorthogonal nanozymes through the encapsulation of hydrophobic transition metal catalysts into the monolayer of water-soluble gold nanoparticles. The activity of these catalysts can be reversibly controlled by binding a supramolecular cucurbit[7]uril 'gate-keeper' onto the monolayer surface, providing a biomimetic control mechanism that mimics the allosteric regulation of enzymes. The potential of this gated nanozyme for use in imaging and therapeutic applications was demonstrated through triggered cleavage of allylcarbamates for pro-fluorophore activation and propargyl groups for prodrug activation inside living cells.
Sigma-Aldrich
ADA, ≥98% (titration)