MilliporeSigma
  • X-ray structure analysis reveals β-turn mimicry by N-amino-imidazolidin-2-ones.
X-ray structure analysis reveals β-turn mimicry by N-amino-imidazolidin-2-ones.
Biopolymers (2015-03-20)
Ngoc-Duc Doan, William D Lubell
ABSTRACT
The conformation of the N-amino-imidazolidin-2-one (Aid) peptidomimetic was investigated using NMR spectroscopy and X-ray crystallography. In solution, the tetrapeptide model p-bromobenzoyl-Aid-l-Phe-N'-iso-propylamide (1) exhibited shielded and solvent exposed amide protons indicative of a turn conformation. In the crystal lattice of 1, four turn conformers were present in the unit cell differing primarily by the ring puckering of the Aid residue. Two pairs of β-turn conformers were observed each possessing an intramolecular ten-member hydrogen bond between the benzamide carbonyl and iso-propylamine NH moieties. The pairs had types II and II' β-turn geometry that differed slightly about the ϕ and ψ torsion angles. Moreover, the X-ray analysis of 1 has been compared with that of the related unsaturated N-amino-imidazolin-2-one (Nai) analogs indicating the influences of ring flattening and substituents on conformation. Insertion of the preorganized Aid structure into biologically active peptides has thus been shown to offer a potentially valuable means for exploring the importance of turn geometry for activity, particularly in drug discovery.
Sigma-Aldrich
Hydroxylamine hydrochloride, JIS special grade, ≥98.0%
Sigma-Aldrich
1,2-Dibromoethane, 98%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
1,2-Dibromoethane, ≥99%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Pyridine, JIS special grade, ≥99.5%
Supelco
1,2-Dibromoethane solution, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%