MilliporeSigma
The effect of phosphate on the nuclease activity of vanadium compounds.
Journal of inorganic biochemistry (2015-05-12)
Nataliya Butenko, José Paulo Pinheiro, José Paulo Da Silva, Ana Isabel Tomaz, Isabel Correia, Vera Ribeiro, João Costa Pessoa, Isabel Cavaco
ABSTRACT
The nuclease activity of VO(acac)2 (1, acac = acetylacetone) and its derivatives VO(hd)2 (2, hd = 3,5-heptanedione), VO(Cl-acac)2 (3, Cl-acac = 3-chloro-2,4-pentanedione), VO(Et-acac)2 (4, Et-acac = 3-ethyl-2,4-pentanedione) and VO(Me-acac)2 (5, Me-acac = 3-methyl-2,4-pentanedione), is studied by agarose gel electrophoresis, UV-visible spectroscopy, cyclic and square wave voltammetry and (51)V NMR. The mechanism is shown to be oxidative and associated with the formation of reactive oxygen species (ROS). Hydrolytic cleavage of the phosphodiester bond is also promoted by 1, but at much slower rate which cannot compete with the oxidative mechanism. The generation of ROS is much higher in the presence of phosphate buffer when compared with organic buffers and this was attributed to the formation of a mixed-ligand complex containing phosphate, (V(IV)O)(V(V)O)(acac)2(HnPO4(n-3)), presenting a quasi-reversible voltammetric behavior. The formation of this species was further observed by Electrospray Ionization Mass Spectrometry (ESI-MS). Phosphate being an essential species in most biological media, the importance of the formation of mixed-ligand species in other vanadium systems is emphasized.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M