- A novel type I receptor serine-threonine kinase predominantly expressed in the adult central nervous system.
A novel type I receptor serine-threonine kinase predominantly expressed in the adult central nervous system.
Receptor serine-threonine kinases (RSTK) mediate inhibitory as well as stimulatory signals for growth and differentiation by binding to members of the transforming growth factor-beta (TGF-beta) superfamily. Over 12 different RSTKs have been isolated so far, displaying wide expression in peripheral tissues and in the nervous system. Here we report the isolation and characterization of a novel type I RSTK termed activin receptor-like kinase-7 (ALK-7) that, unlike other members of this receptor family, is predominantly expressed in the adult central nervous system. The ALK-7 gene encodes a 55-kDa cell-surface protein that exhibits up to 78% amino acid sequence identity in the kinase domain to previously isolated type I receptors for TGF-beta and activin. In the extracellular domain, however, ALK-7 is more divergent, displaying comparable similarities with all members of the ALK subfamily. RNase protection and in situ hybridization studies demonstrated a highly specific mRNA distribution restricted to neurons in several regions of the adult rat central nervous system, including cerebellum, hippocampus, and nuclei of the brainstem. Receptor reconstitution and cross-linking experiments indicated that ALK-7 can form complexes with type II RSTKs for TGF-beta and activin in a ligand-dependent manner, although direct binding of ALK-7 to ligand in these complexes could not be demonstrated. The specific expression pattern of ALK-7, restricted to the postnatal central nervous system, indicates that this receptor may play an important role in the maturation and maintenance of several neuronal subpopulations.