MilliporeSigma
  • Detection of Dinophysistoxin-1 in Clonal Culture of Marine Dinoflagellate Prorocentrum foraminosum (Faust M.A., 1993) from the Sea of Japan.
Detection of Dinophysistoxin-1 in Clonal Culture of Marine Dinoflagellate Prorocentrum foraminosum (Faust M.A., 1993) from the Sea of Japan.
Toxins (2015-10-02)
Polina A Kameneva, Kseniya V Efimova, Viacheslav G Rybin, Tatiana Y Orlova
ABSTRACT
For the first time the presence of dinophysistoxin-1 (DTX-1) in a culture of Prorocentrum foraminosum was revealed in cells and in the culture medium. The clone was isolated from coastal waters of the Sea of Japan and identified by molecular analyses of SSU and D1/D2 regions of LSU rDNA. The concentration of DTX-1 in cells was 8.4 ± 2.5 pg/cell and, in cell-free media, 27.9 ± 14.7 µg/L. The toxin presence was confirmed by HPLC with high-resolution tandem mass-spectrometry.
Sigma-Aldrich
18-Crown-6, 99%
Sigma-Aldrich
18-Crown-6, ≥99.0%
Sigma-Aldrich
4-Bromomethyl-7-methoxycoumarin, 97%
Sigma-Aldrich
18-Crown-6 solution, 1.0 M in THF
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Potassium carbonate, puriss., meets analytical specification of Ph. Helv., anhydrous, granulated, 99-101% (calc. to the dried substance)