MilliporeSigma
  • Tat/HA2 Peptides Conjugated AuNR@pNIPAAm as a Photosensitizer Carrier for Near Infrared Triggered Photodynamic Therapy.
Tat/HA2 Peptides Conjugated AuNR@pNIPAAm as a Photosensitizer Carrier for Near Infrared Triggered Photodynamic Therapy.
Molecular pharmaceutics (2015-06-03)
Shefang Ye, Ning Kang, Min Chen, Caiding Wang, Tianxiao Wang, Yarun Wang, Yongliang Liu, Donghui Li, Lei Ren
ABSTRACT
To achieve an efficiency of intracellular photosensitizers (PSs) delivery and efficacy of photodynamic therapy, we have developed a novel class of PS formulation for encapsulating sulfonated aluminum phthalocyanine (AlPcS4) by taking advantage of the membrane-disruptive peptides Tat/HA2 and the photothermally triggered delivery system using AuNR@pNIPAAm. The coordinated effects of cell penetrating peptide Tat and fusogenic peptide HA2 could enhance the efficient cellular internalization and endo/lysosome escape of PSs delivery systems. Singlet oxygen generation was inhibited due to the reaction between loaded AlPcS4 and Au nanorods, which indicated that the AlPcS4-loaded, AuNR@pNIPAAm delivery system might be nonphototoxic in the circulatory system. However, this PSs-loaded nanosystem became highly phototoxic as it underwent the near-infrared irradiation by using the combined lights of 808 and 680 nm. Upon irradiation, the Tat/HA2 conjugated AuNR@pNIPAAm-Pc elicited an active photodynamic response against the cancer cells, leading to effective cells killing via mitochondria-associated apoptotic pathway. This study also demonstrated improved PDT therapeutic efficacy after intravenous administration of Tat/HA2-AuNR@pNIPAAm-Pc and the subsequent lights irradiations in tumor-bearing mice. We describe here a strategy for enhanced photodynamic eradication of solid tumors by endo/lysosomal escape and highlight the great promise of peptide-based nanocarriers used for cancer therapy.
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Propidium iodide solution