MilliporeSigma
  • Hemocytes of Rhipicephalus sanguineus (Acari: Ixodidae): Characterization, Population Abundance, and Ultrastructural Changes Following Challenge with Leishmania infantum.
Hemocytes of Rhipicephalus sanguineus (Acari: Ixodidae): Characterization, Population Abundance, and Ultrastructural Changes Following Challenge with Leishmania infantum.
Journal of medical entomology (2015-09-04)
A P S Feitosa, L C Alves, M M Chaves, D L Veras, E M Silva, A S S Aliança, I R S França, G G A Gonçalves, J L Lima-Filho, F A Brayner
ABSTRACT
Few studies have examined the cellular immune response of ticks, and further research on the characterization of the hemocytes of ticks is required, particularly on those of Rhipicephalus sanguineus (Latreille) because of the medical and veterinary importance of this tick. The aims of this study were to characterize the morphology and the ultrastructure of the different types of hemocytes of adult R. sanguineus and to determine the population abundance and the ultrastructural changes in the hemocytes of ticks infected with Leishmania infantum. The hemocytes were characterized through light and transmission electron microscopy. Within the variability of circulating cells in the hemolymph of adult R. sanguineus, five cell types were identified, which were the prohemocytes, plasmatocytes, granulocytes, spherulocytes, and adipohemocytes. The prohemocytes were the smallest cells found in the hemolymph. The plasmatocytes had polymorphic morphology with vesicles and cytoplasmic projections. The granulocytes had an elliptical shape with the cytoplasm filled with granules of different sizes and electrodensities. The spherulocytes were characterized by several spherules of uniform shapes and sizes that filled the entire cytoplasm, whereas the adipohemocytes had an irregular shape with multiple lipid inclusions that occupied almost the entire cytoplasmic space. The total counts of the hemocyte population increased in the group that was infected with L. infantum. Among the different cell types, the numbers increased and the ultrastructural changes occurred in the granulocytes and the plasmatocytes in the infected group of ticks.
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF