MilliporeSigma
  • Significantly improved solvent tolerance of Escherichia coli by global transcription machinery engineering.
Significantly improved solvent tolerance of Escherichia coli by global transcription machinery engineering.
Microbial cell factories (2015-11-07)
Fa Zhang, Xiaohong Qian, Haiming Si, Guochao Xu, Ruizhi Han, Ye Ni
ABSTRACT
Escherichia coli has emerged as a promising platform microorganism to produce biofuels and fine chemicals of industrial interests. Certain obstacles however remain to be overcome, among which organic-solvent tolerance is a crucial one. We used global transcription machinery engineering (gTME) to improve the organic-solvent tolerance (OST) of E. coli JM109. A mutant library of σ(70) encoded by rpoD was screened under cyclohexane pressure. E. coli JM109 strain harboring σ(70) mutant C9 was identified with capability of tolerating 69 % cyclohexane. The rpoD mutant contains three amino-acid substitutes and a stop-codon mutation, resulting a truncated sequence containing regions σ(1.1) and σ(1.2). Total protein difference produced by E. coli JM109 strain harboring C9 was examined with 2D-PAGE, and 204 high-abundant proteins showed over twofold variation under different solvent stress. Our results show that several genes (gapA, sdhB, pepB and dppA) play critical roles in enhanced solvent tolerance of E. coli, mainly involving in maintaining higher intracellular energy level under solvent stress. Global transcription machinery engineering is therefore a feasible and efficient approach for engineering strain with enhanced OST-phenotype.
Sigma-Aldrich
Cyclohexane, JIS special grade
Sigma-Aldrich
Cyclohexane, suitable for HPLC
Sigma-Aldrich
Cyclohexane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%