MilliporeSigma
  • Single-cell genomics-based analysis of virus-host interactions in marine surface bacterioplankton.
Single-cell genomics-based analysis of virus-host interactions in marine surface bacterioplankton.
The ISME journal (2015-04-08)
Jessica M Labonté, Brandon K Swan, Bonnie Poulos, Haiwei Luo, Sergey Koren, Steven J Hallam, Matthew B Sullivan, Tanja Woyke, K Eric Wommack, Ramunas Stepanauskas
ABSTRACT
Viral infections dynamically alter the composition and metabolic potential of marine microbial communities and the evolutionary trajectories of host populations with resulting feedback on biogeochemical cycles. It is quite possible that all microbial populations in the ocean are impacted by viral infections. Our knowledge of virus-host relationships, however, has been limited to a minute fraction of cultivated host groups. Here, we utilized single-cell sequencing to obtain genomic blueprints of viruses inside or attached to individual bacterial and archaeal cells captured in their native environment, circumventing the need for host and virus cultivation. A combination of comparative genomics, metagenomic fragment recruitment, sequence anomalies and irregularities in sequence coverage depth and genome recovery were utilized to detect viruses and to decipher modes of virus-host interactions. Members of all three tailed phage families were identified in 20 out of 58 phylogenetically and geographically diverse single amplified genomes (SAGs) of marine bacteria and archaea. At least four phage-host interactions had the characteristics of late lytic infections, all of which were found in metabolically active cells. One virus had genetic potential for lysogeny. Our findings include first known viruses of Thaumarchaeota, Marinimicrobia, Verrucomicrobia and Gammaproteobacteria clusters SAR86 and SAR92. Viruses were also found in SAGs of Alphaproteobacteria and Bacteroidetes. A high fragment recruitment of viral metagenomic reads confirmed that most of the SAG-associated viruses are abundant in the ocean. Our study demonstrates that single-cell genomics, in conjunction with sequence-based computational tools, enable in situ, cultivation-independent insights into host-virus interactions in complex microbial communities.
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)