MilliporeSigma
  • TRPV1 sensitization mediates postinflammatory visceral pain following acute colitis.
TRPV1 sensitization mediates postinflammatory visceral pain following acute colitis.
American journal of physiology. Gastrointestinal and liver physiology (2015-05-30)
Tamia K Lapointe, Lilian Basso, Mircea C Iftinca, Robyn Flynn, Kevin Chapman, Gilles Dietrich, Nathalie Vergnolle, Christophe Altier
ABSTRACT
Quiescent phases of inflammatory bowel disease (IBD) are often accompanied by chronic abdominal pain. Although the transient receptor potential vanilloid 1 (TRPV1) ion channel has been postulated as an important mediator of visceral hypersensitivity, its functional role in postinflammatory pain remains elusive. This study aimed at establishing the role of TRPV1 in the peripheral sensitization underlying chronic visceral pain in the context of colitis. Wild-type and TRPV1-deficient mice were separated into three groups (control, acute colitis, and recovery), and experimental colitis was induced by oral administration of dextran sulfate sodium (DSS). Recovery mice showed increased chemically and mechanically evoked visceral hypersensitivity 5 wk post-DSS discontinuation, at which point inflammation had completely resolved. Significant changes in nonevoked pain-related behaviors could also be observed in these animals, indicative of persistent discomfort. These behavioral changes correlated with elevated colonic levels of substance P (SP) and TRPV1 in recovery mice, thus leading to the hypothesis that SP could sensitize TRPV1 function. In vitro experiments revealed that prolonged exposure to SP could indeed sensitize capsaicin-evoked currents in both cultured neurons and TRPV1-transfected human embryonic kidney (HEK) cells, a mechanism that involved TRPV1 ubiquitination and subsequent accumulation at the plasma membrane. Importantly, although TRPV1-deficient animals experienced similar disease severity and pain as wild-type mice in the acute phase of colitis, TRPV1 deletion prevented the development of postinflammatory visceral hypersensitivity and pain-associated behaviors. Collectively, our results suggest that chronic exposure of colon-innervating primary afferents to SP could sensitize TRPV1 and thus participate in the establishment of persistent abdominal pain following acute inflammation.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride solution, 1 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Pricing and availability is not currently available. Contact Technical Service
SAFC
HEPES
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available. Contact Technical Service