MilliporeSigma
  • Efficient recovery of undifferentiated human embryonic stem cell cryopreserved with hydroxyethyl starch, dimethyl sulphoxide and serum replacement.
Efficient recovery of undifferentiated human embryonic stem cell cryopreserved with hydroxyethyl starch, dimethyl sulphoxide and serum replacement.
Cryobiology (2015-02-03)
Maristela Delgado Orellana, Gil Cunha De Santis, Kuruvilla Joseph Abraham, Aparecida Maria Fontes, Danielle Aparecida Rosa Magalhães, Viviane de Cássia Oliveira, Everton de Brito Oliveira Costa, Patrícia Vianna Bonini Palma, Dimas Tadeu Covas
ABSTRACT
The therapeutic use of human embryonic stem cells (hESCs) is dependent on an efficient cryopreservation protocol for long-term storage. The aim of this study was to determine whether the combination of three cryoprotecting reagents using two freezing systems might improve hESC recovery rates with maintenance of hESC pluripotency properties for potential cell therapy application. Recovery rates of hESC colonies which were frozen in three cryoprotective solutions: Me2SO/HES/SR medium, Defined-medium® and Me2SO/SFB in medium solution were evaluated in ultra-slow programmable freezing system (USPF) and a slow-rate freezing system (SRF). The hESC pluripotency properties after freezing-thawing were evaluated. We estimated the distribution frequency of survival colonies and observed that independent of the freezing system used (USPF or SRF) the best results were obtained with Me2SO/HES/SR as cryopreservation medium. We showed a significant hESC recovery colonies rate after thawing in Me2SO/HES/SR medium were 3.88 and 2.9 in USPF and SRF, respectively. The recovery colonies rate with Defined-medium® were 1.05 and 1.07 however in classical Me2SO medium were 0.5 and 0.86 in USPF and SRF, respectively. We showed significant difference between Me2SO/HES/SR medium×Defined-medium® and between Me2SO/HES/SR medium×Me2SO medium, for two cryopreservation systems (P<0.05). We developed an in house protocol using the combination of Me2SO/HES/SR medium and ultra-slow programmable freezing system which resulted in hESC colonies that remain undifferentiated, maintain their in vitro and in vivo pluripotency properties and genetic stability. This approach may be suitable for cell therapy studies.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, 99.5-100.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
SAFC
Glycine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis