MilliporeSigma
  • Fibrotic Effects of Arecoline N-Oxide in Oral Potentially Malignant Disorders.
Fibrotic Effects of Arecoline N-Oxide in Oral Potentially Malignant Disorders.
Journal of agricultural and food chemistry (2015-06-11)
Tzer-Min Kuo, Shun-Yuan Luo, Shang-Lun Chiang, Kun-Tu Yeh, Hui-Ting Hsu, Cheng-Tien Wu, Chi-Yu Lu, Ming-Hsui Tsai, Jan-Gowth Chang, Ying-Chin Ko
ABSTRACT
The metabolites of environmental chemicals play key roles in carcinogenesis. Areca nut is strongly associated with the development of oral potentially malignant disorders (OPMD) or cancer. The main alkaloid in the areca nut is arecoline, which is highly cytotoxic and genotoxic. Arecoline N-oxide, a metabolite of areca nut alkaloids, which has been identified in animal urine, has been shown to induce mutagenicity in bacteria. In this study, it was found that its protein adduct could be detected in oral keratinocytes treated with areca nut extract. Increased collagen expression and severity of squamous hyperplasia were observed in arecoline N-oxide treated mice. In cultured oral fibroblasts, arecoline N-oxide showed stronger effects on the increase of fibrotic related genes including TGF-beta1, S100A4, MMP-9, IL-6, and fibronectin and a decrease of E-cadherin as compared with arecoline. Finally, arecoline N-oxide stimulation effectively increased the DNA damage marker, gamma-H2A.X, both in vitro and in vivo. Taken together, these results indicate that arecoline N-oxide shows a high potential for the induction of OPMD.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M