MilliporeSigma
  • Dissociation of heroin-induced emotional dysfunction from psychomotor activation and physical dependence among inbred mouse strains.
Dissociation of heroin-induced emotional dysfunction from psychomotor activation and physical dependence among inbred mouse strains.
Psychopharmacology (2014-12-09)
G Ayranci, K Befort, L Lalanne, B L Kieffer, P-E Lutz
ABSTRACT
Opiate addiction is a brain disorder emerging through repeated intoxication and withdrawal episodes. Epidemiological studies also indicate that chronic exposure to opiates may lead in susceptible individuals to the emergence of depressive symptoms, strongly contributing to the severity and chronicity of addiction. We recently established a mouse model of heroin abstinence, characterized by the development of depressive-like behaviors following chronic heroin exposure. While genetic factors regulating immediate behavioral responses to opiates have been largely investigated, little is known about their contribution to long-term emotional regulation during abstinence. Here, we compared locomotor stimulation and physical dependence induced by heroin exposure, as well as emotional dysfunction following abstinence, across mice strains with distinct genetic backgrounds. Mice from three inbred strains (C57BL/6J, Balb/cByJ, and 129S2/SvPas) were exposed to an escalating chronic heroin regimen (10-50 mg/kg). Independent cohorts were used to assess drug-induced locomotor activity during chronic treatment, naloxone-precipitated withdrawal at the end of chronic treatment, and emotional-like responses after a 4-week abstinence period. Distinct behavioral profiles were observed across strains during heroin treatment, with no physical dependence and low locomotor stimulation in 129S2/SvPas. In addition, different behavioral impairments developed during abstinence across the three strains, with increased despair-like behavior in 129S2/SvPas and Balb/cByJ, and low sociability in 129S2/SvPas and C57BL/6J. Our results indicate that depressive-like behaviors emerge during heroin abstinence, whatever the severity of immediate behavioral responses to the drug. In addition, inbred mouse strains will allow studying several aspects of mood-related deficits associated with addiction.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
Sodium chloride solution, 5 M