MilliporeSigma
  • Superior antitumor activity of trastuzumab combined with capecitabine plus oxaliplatin in a human epidermal growth factor receptor 2-positive human gastric cancer xenograft model.
Superior antitumor activity of trastuzumab combined with capecitabine plus oxaliplatin in a human epidermal growth factor receptor 2-positive human gastric cancer xenograft model.
Molecular and clinical oncology (2015-12-02)
Suguru Harada, Mieko Yanagisawa, Saori Kaneko, Keigo Yorozu, Kaname Yamamoto, Yoichiro Moriya, Naoki Harada
ABSTRACT
In the treatment of human epidermal growth factor receptor 2 (HER2)-positive advanced gastric or gastroesophageal junction cancer, it has been reported that the combination of trastuzumab with capecitabine plus cisplatin, or with 5-fluorouracil (5-FU) plus cisplatin, significantly increased overall survival compared with chemotherapy alone (ToGA trial). In addition, adjuvant therapy with capecitabine plus oxaliplatin (XELOX) improved the survival of patients who received curative D2 gastrectomy (CLASSIC trial). However, the efficacy of the combination of trastuzumab with XELOX for patients with HER2-positive gastric cancer remains unknown. The aim of this study, was to investigate the efficacy of the combination of trastuzumab with XELOX in a HER2-positive human gastric cancer xenograft model. Combination treatment with these three agents (trastuzumab 20 mg/kg, capecitabine 359 mg/kg and oxaliplatin 10 mg/kg), was found to exhibit a significantly stronger antitumor activity in NCI-N87 xenografts compared with either trastuzumab or XELOX alone. In this model, treatment with trastuzumab alone or trastuzumab plus oxaliplatin enhanced the expression of thymidine phosphorylase (TP), a key enzyme in the generation of 5-FU from capecitabine in tumor tissues. In