MilliporeSigma
  • p24A, a type I transmembrane protein, controls ARF1-dependent resensitization of protease-activated receptor-2 by influence on receptor trafficking.
p24A, a type I transmembrane protein, controls ARF1-dependent resensitization of protease-activated receptor-2 by influence on receptor trafficking.
The Journal of biological chemistry (2007-08-19)
Weibo Luo, Yingfei Wang, Georg Reiser
ABSTRACT
Protease-activated receptor-2 (PAR-2), the second member of the G protein-coupled PAR family, is irreversibly activated by trypsin or tryptase and then targeted to lysosomes for degradation. Intracellular presynthesized receptors stored at the Golgi apparatus repopulate the cell surface after trypsin stimulation, thereby leading to rapid resensitization to trypsin signaling. However, the molecular mechanisms of the exocytic trafficking of PAR-2 from the Golgi apparatus to the plasma membrane remain largely unclear. Here we show that p24A, a type I transmembrane protein, which is a crucial constituent of the Golgi apparatus, associates with PAR-2 at the Golgi apparatus. The protein interaction occurs between the N-terminal region of p24A (residues 1-105; p24A-GL (GOLD domain with a small linker)) and the second extracellular loop of PAR-2. After receptor activation, PAR-2 dissociates from p24A. Importantly, we found that ADP-ribosylation factor 1 regulated the dissociation process and initiated PAR-2 trafficking to the plasma membrane. Conversely, overexpression of the fragment p24A-GL, but not other mutants containing the functional coiled-coil domain of p24A, arrested PAR-2 at the Golgi apparatus and inhibited receptor trafficking to the plasma membrane, which consequently prevented resensitization of PAR-2. These findings identify a new function of p24A as a regulator of signal-dependent trafficking that regulates the life cycle of PAR-2, Thus, we reveal a new molecular mechanism underlying resensitization of PAR-2.