MilliporeSigma
  • Pipette vial dispersive liquid-liquid microextraction combined with high-performance liquid chromatography for the determination of benzoylurea insecticide in fruit juice.
Pipette vial dispersive liquid-liquid microextraction combined with high-performance liquid chromatography for the determination of benzoylurea insecticide in fruit juice.
Journal of separation science (2015-11-04)
Xuefei Xi, Miyi Yang, Ganni Shen, Xiaoling Wu, Runhua Lu, Wenfeng Zhou, Sanbing Zhang, Haixiang Gao
ABSTRACT
A simple, sensitive, and efficient method of using a pipette vial to perform dispersive liquid-liquid microextraction based on the solidification of floating organic droplets was coupled with high-performance liquid chromatography (HPLC) and a diode array detector for the preconcentration and analysis of four benzoylurea insecticides in fruit juice. In this method, 1-dodecanol was used as an extractant, and a snipped pipette was used as an experimental vial to simplify the procedure of collecting and separating solidified extractant. The experimental parameters were optimized using a Plackett-Burman design and one-factor-at-a-time method. Under the optimal conditions in the water model, the limits of detection for analytes varied from 0.03 to 0.28 μg/L, and the enrichment factors ranged from 147 to 206. Linearity was achieved for diflubenzuron and flufenoxuron in a range of 0.5-500 μg/L, for hexaflumuron in a range of 1-500 μg/L, and for triflumuron in a range of 5-500 μg/L. The correlation coefficients for the analytes ranged from 0.9986 to 0.9994 with recoveries of 91.4-110.9%. Finally, the developed technique was successfully applied to fruit juice samples with acceptable results. The relative standard deviations of the analytes at two spiking levels (50 and 200 μg/L) varied between 0.2 and 4.5%.
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%
Pricing and availability is not currently available.