MilliporeSigma
  • Development of a genomic DNA reference material panel for myotonic dystrophy type 1 (DM1) genetic testing.
Development of a genomic DNA reference material panel for myotonic dystrophy type 1 (DM1) genetic testing.
The Journal of molecular diagnostics : JMD (2013-05-18)
Lisa Kalman, Jack Tarleton, Monica Hitch, Madhuri Hegde, Nick Hjelm, Elizabeth Berry-Kravis, Lili Zhou, James E Hilbert, Elizabeth A Luebbe, Richard T Moxley, Lorraine Toji
ABSTRACT
Myotonic dystrophy type 1 (DM1) is caused by expansion of a CTG triplet repeat in the 3' untranslated region of the DMPK gene that encodes a serine-threonine kinase. Patients with larger repeats tend to have a more severe phenotype. Clinical laboratories require reference and quality control materials for DM1 diagnostic and carrier genetic testing. Well-characterized reference materials are not available. To address this need, the Centers for Disease Control and Prevention-based Genetic Testing Reference Material Coordination Program, in collaboration with members of the genetic testing community, the National Registry of Myotonic Dystrophy and Facioscapulohumeral Muscular Dystrophy Patients and Family Members, and the Coriell Cell Repositories, has established and characterized cell lines from patients with DM1 to create a reference material panel. The CTG repeats in genomic DNA samples from 10 DM1 cell lines were characterized in three clinical genetic testing laboratories using PCR and Southern blot analysis. DMPK alleles in the samples cover four of five DM1 clinical categories: normal (5 to 34 repeats), mild (50 to 100 repeats), classical (101 to 1000 repeats), and congenital (>1000 repeats). We did not identify or establish Coriell cell lines in the premutation range (35 to 49 repeats). These samples are publicly available for quality control, proficiency testing, test development, and research and should help improve the accuracy of DM1 testing.