MilliporeSigma
  • Effect of Aspect Ratio on Field Emission Properties of ZnO Nanorod Arrays.
Effect of Aspect Ratio on Field Emission Properties of ZnO Nanorod Arrays.
Nanoscale research letters (2008-01-01)
Xuemin Qian, Huibiao Liu, Yanbing Guo, Yinglin Song, Yuliang Li
ABSTRACT
ZnO nanorod arrays are prepared on a silicon wafer through a multi-step hydrothermal process. The aspect ratios and densities of the ZnO nanorod arrays are controlled by adjusting the reaction times and concentrations of solution. The investigation of field emission properties of ZnO nanorod arrays revealed a strong dependency on the aspect ratio and their density. The aspect ratio and spacing of ZnO nanorod arrays are 39 and 167 nm (sample C), respectively, to exhibit the best field emission properties. The turn-on field and threshold field of the nanorod arrays are 3.83 V/μm and 5.65 V/μm, respectively. Importantly, the sample C shows a highest enhancement of factorβ, which is 2612. The result shows that an optimum density and aspect ratio of ZnO nanorod arrays have high efficiency of field emission.
Sigma-Aldrich
Zinc nitrate hexahydrate, purum p.a., crystallized, ≥99.0% (KT)
Pricing and availability is not currently available.
Sigma-Aldrich
Zinc nitrate hexahydrate, reagent grade, 98%
Pricing and availability is not currently available.