Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl palmitate, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl oleate, technical grade, 70%
SKU
Pack Size
Availability
Price
Quantity
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl stearate, ≥96%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl stearate, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl linoleate, technical, ≥65% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl oleate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl linoleate, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl stearate, ≥99% (capillary GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl stearate, ~99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl oleate, natural, ≥85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%