MilliporeSigma
  • Differential Gene Expression Profiles and Selected Cytokine Protein Analysis of Mediastinal Lymph Nodes of Horses with Chronic Recurrent Airway Obstruction (RAO) Support an Interleukin-17 Immune Response.
Differential Gene Expression Profiles and Selected Cytokine Protein Analysis of Mediastinal Lymph Nodes of Horses with Chronic Recurrent Airway Obstruction (RAO) Support an Interleukin-17 Immune Response.
PloS one (2015-11-13)
Alexandra Korn, Donald Miller, Lynn Dong, Elizabeth Louise Buckles, Bettina Wagner, Dorothy Marie Ainsworth
ABSTRACT
Recurrent airway obstruction (RAO) is a pulmonary inflammatory condition that afflicts certain mature horses exposed to organic dust particulates in hay. Its clinical and pathological features, manifested by reversible bronchoconstriction, excessive mucus production and airway neutrophilia, resemble the pulmonary alterations that occur in agricultural workers with occupational asthma. The immunological basis of RAO remains uncertain although its chronicity, its localization to a mucosal surface and its domination by a neutrophilic, non-septic inflammatory response, suggest involvement of Interleukin-17 (IL-17). We examined global gene expression profiles in mediastinal (pulmonary-draining) lymph nodes isolated from RAO-affected and control horses. Differential expression of > 200 genes, coupled with network analysis, supports an IL-17 response centered about NF-κB. Immunohistochemical analysis of mediastinal lymph node sections demonstrated increased IL-17 staining intensity in diseased horses. This result, along with the finding of increased IL-17 concentrations in lymph node homogenates from RAO-affected horses (P = 0.1) and a down-regulation of IL-4 gene and protein expression, provides additional evidence of the involvement of IL-17 in the chronic stages of RAO. Additional investigations are needed to ascertain the cellular source of IL-17 in this equine model of occupational asthma. Understanding the immunopathogenesis of this disorder likely will enhance the development of therapeutic interventions beneficial to human and animal pulmonary health.