MilliporeSigma
  • Array-CGH analysis in patients with syndromic and non-syndromic XY gonadal dysgenesis: evaluation of array CGH as diagnostic tool and search for new candidate loci.
Array-CGH analysis in patients with syndromic and non-syndromic XY gonadal dysgenesis: evaluation of array CGH as diagnostic tool and search for new candidate loci.
Human reproduction (Oxford, England) (2010-08-06)
S Ledig, O Hiort, G Scherer, M Hoffmann, G Wolff, S Morlot, A Kuechler, P Wieacker
ABSTRACT
XY gonadal dysgenesis (XY-GD) is a heterogeneous disorder characterized by failure of testicular development despite a normal male karyotype. Non-syndromic and syndromic forms can be delineated. Currently, only a minority of cases can be explained by gene mutations. The aim of this study was to detect microdeletions and duplications by using high-resolution Agilent oligonucleotide arrays in a cohort of 87 patients with syndromic or non-syndromic 46,XY-GD. In 26 patients, we identified gains or losses in regions including genes involved in XY-GD (DMRT1, SOX9, DAX1) or in regions, which have not been described as polymorphic copy number variants (CNVs). This study shows that array comparative genomic hybridization (CGH) analysis is a useful tool for the molecular diagnosis of XY-GD as well as for the identification of potential candidate genes involved in male sexual development.