MilliporeSigma
  • T cell reconstitution in allogeneic haematopoietic stem cell transplantation: prognostic significance of plasma interleukin-7.
T cell reconstitution in allogeneic haematopoietic stem cell transplantation: prognostic significance of plasma interleukin-7.
Scandinavian journal of immunology (2014-09-30)
K Kielsen, K K Jordan, H H Uhlving, P L Pontoppidan, Z Shamim, M Ifversen, C Heilmann, C H Nielsen, H Sengeløv, L P Ryder, K G Müller
ABSTRACT
Infections and acute graft-versus-host disease (aGVHD) are major causes of treatment-related mortality and morbidity following allogeneic haematopoietic stem cell transplantation (HSCT). Both complications depend on reconstitution of the T-lymphocyte population based on donor T cells. Although it is well established that Interleukin-7 (IL-7) is a cytokine essential for de novo T cell development in the thymus and homoeostatic peripheral expansion of T cells, associations between circulating levels of IL-7 and T cell reconstitution following HSCT have not been investigated previously. We prospectively measured IL-7 levels in 81 patients undergoing myeloablative HSCT with either sibling donor or an unrelated donor. Plasma IL-7 levels peaked at day +7 post-transplant (1.3-82.4 pg/ml), at the time of maximal lymphopaenia. In multivariate analysis, peak levels of IL-7 were significantly higher in patients treated with anti-thymocyte globulin (ATG) compared with those not treated with ATG (P = 0.0079). IL-7 levels at day +7 were negatively associated with T cell counts at day +30 to +60 (at day +60: CD3(+) : β = -10.6 × 10(6) cells/l, P = 0.0030; CD8(+) : β = -8.4 × 10(6) cells/l, P = 0.061; CD4(+) : β = -2.1 × 10(6) cells/l, P = 0.062) in multivariate analyses. In adults, high IL-7 levels were associated with increased risk of grade II-IV aGVHD (OR = 5.4, P = 0.036) and reduced overall survival (P = 0.046). The present data indicate that high plasma levels of IL-7 in the early post-transplant period are predictive for slow T cell reconstitution, increased risk of aGVHD and increased mortality following HSCT.
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)