MilliporeSigma
  • Metastasis associated in colon cancer 1 predicts poor outcomes in patients with breast cancer.
Metastasis associated in colon cancer 1 predicts poor outcomes in patients with breast cancer.
Analytical and quantitative cytopathology and histopathology (2015-06-13)
Ga-Eon Kim, Ji Shin Lee, Min Ho Park, Jung Han Yoon
ABSTRACT
To investigate whether metastasis associated in colon cancer 1 (MACC1) expression is a prognostic marker in breast cancer and to demonstrate the potential correlation between MACC1 and mitogen-activated protein kinase (MAPK) expression. Immunohistochemical staining with anti-MACC1 and phospho-p44/42 MAPK antibodies was performed in 198 invasive breast carcinomas using tissue microarray. Expression of MACC1 was detected in 109 (55.1%) of 198 invasive breast carcinomas. MACC1 expression was significantly higher in the metastatic relapse-positive group (87.8%, 36/41) than in the metastatic relapse-negative group (46.5%, 73/157) (p < 0.001). MACC1 expression was significantly correlated with phospho-p44/42 MAPK expression (p < 0.05). On univariate analysis a significant association was observed between MACC1 expression and decreased disease-free survival (p < 0.001) and overall survival (p = 0.001). On multivariate analysis MACC1 expression was one of the statistically significant independent risk factors for disease-free survival (p = 0.001). MACC1 may serve as a new parameter for the prognostic prediction in patients with invasive breast carcinoma. MACC1 is likely to be involved in the regulation of MAPK cascades in invasive breast carcinoma.