MilliporeSigma
  • Hsp74, a potential bladder cancer marker, has direct interaction with keratin 1.
Hsp74, a potential bladder cancer marker, has direct interaction with keratin 1.
Journal of immunology research (2014-07-23)
Ling Chen, YaRong Wang, Le Zhao, Wei Chen, Chunhui Dong, Xinhan Zhao, Xu Li
ABSTRACT
Early diagnosis and prognosis monitoring are very important for the survival of patients with bladder cancer. To identify candidate biomarkers of bladder cancer, we used a combination of techniques including 2-DE, co-IP, western blot, LC-MS/MS, and immunohistochemistry. Hsp74 was identified with high expression in bladder cancer. The cellular location of expression products of gene Hsp74 showed that they were distributed into cytoplasm and keratin 1 was found to be associated with Hsp74. The results provide a new idea to understand the molecular basis of bladder cancer progression and pinpoint new potential molecular target for early diagnosis and therapeutic monitoring of bladder cancer.