MilliporeSigma
IFIH1-GCA-KCNH7 locus: influence on multiple sclerosis risk.
European journal of human genetics : EJHG (2008-02-21)
Alfonso Martínez, José Luis Santiago, M Carmen Cénit, Virginia de Las Heras, Hermenegildo de la Calle, Miguel Fernández-Arquero, Rafael Arroyo, Emilio G de la Concha, Elena Urcelay
ABSTRACT
A recent genome-wide scan of nonsynonymous SNPs and ulterior validation in case-control and family analyses evidenced a susceptibility locus for type 1 diabetes (T1D) on chromosome 2q24.3. We aimed at testing the effect of this locus in other autoimmune diseases with complex genetic background, such as multiple sclerosis (MS). Four SNPs along the locus, rs13422767, rs2111485, rs1990760 and rs2068330, were genotyped using TaqMan MGB chemistry in 311 T1D and 412 MS patients and 535 ethnically matched healthy controls. The previously reported association of this locus was found for the first time in MS (rs2068330, G vs C: P=0.001; OR (95% CI)=0.73 (0.6-0.88)) and a trend for replication was observed in our Spanish diabetic cohort. Therefore, genes included in this locus - IFIH1 interferon induced helicase, GCA grancalcin or the potassium channel KCNH7 - are potential candidates implicated in the pathogenesis of these autoimmune diseases, although strong linkage disequilibrium in the region hampered further localization of the etiologic gene.