MilliporeSigma
  • Syntheses of Sceptrins and Nakamuric Acid and Insights into the Biosyntheses of Pyrrole-Imidazole Dimers.
Syntheses of Sceptrins and Nakamuric Acid and Insights into the Biosyntheses of Pyrrole-Imidazole Dimers.
Organic chemistry frontiers : an international journal of organic chemistry (2015-09-04)
Xiaolei Wang, Yang Gao, Zhiqiang Ma, Rodrigo A Rodriguez, Zhi-Xiang Yu, Chuo Chen
ABSTRACT
Sceptrins and nakamuric acid are structurally unique antibiotics isolated from marine sponges. Recent studies suggest that the biosynthesis of these dimeric pyrrole-imidazole alkaloids involves a single-electron transfer (SET)-promoted [2+2] cycloaddition to form their cyclobutane core skeletons. We describe herein the biomimetic syntheses of racemic sceptrin and nakamuric acid. We also report the asymmetric syntheses of sceptrin, bromosceptrin, and dibromosceptrin in their natural enantiomeric form. We further provide mechanistic insights into the pathway selectivity of the SET-promoted [2+2] and [4+2] cycloadditions that lead to the divergent formation of the sceptrin and ageliferin core skeletons. Both the [2+2] and [4+2] cycloadditions are stepwise reactions, with the [2+2] pathway kinetically and thermodynamically favored over the [4+2] pathway. For the [2+2] cycloaddition, the dimerization of pyrrole-imidazole monomers is rate-limiting, whereas for the [4+2] cycloaddition, the cyclization is the slowest step.