MilliporeSigma
  • Interleukin-16 polymorphism is associated with an increased risk of glioma.
Interleukin-16 polymorphism is associated with an increased risk of glioma.
Genetic testing and molecular biomarkers (2014-08-29)
Qi-Sheng Luo, Jun-Li Wang, Yuan-Yang Deng, Hua-Dong Huang, Huang-De Fu, Chuan-Yu Li, Hai-Neng Huang
ABSTRACT
Previous studies have shown that interleukin (IL)-16 is overexpressed in human and rat gliomas. Potential links between IL-16 polymorphisms and glioma risk are currently unclear. The aim of this study was to investigate the association between IL-16 polymorphisms and glioma risk. We examined IL-16 gene polymorphisms (i.e., rs 4778889, rs 11556218, and rs 4072111) in 216 patients with glioma and 275 controls in a Chinese population. Genotypes were determined using a polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP) assay. Odds ratios (OR) and their corresponding 95% confidence intervals (CI) were used to evaluate the effect of the IL-16 polymorphisms on glioma risk. The rs 11556218TG genotype is associated with an increased risk of glioma compared with the TT genotype (OR=1.76; 95% CI, 1.22-2.54; p=0.002). Similarly, the rs 11556218G allele is associated with an increased risk of glioma compared with the T allele (OR=1.41; 95% CI, 1.06-1.87; p=0.017). However, no significant association was observed between the IL-16 rs 4778889 and rs 4072111 polymorphisms and the risk of glioma. These findings suggest that the IL-16 rs 11556218 polymorphism may be used as a susceptibility marker for glioma.