MilliporeSigma
  • IFT27, encoding a small GTPase component of IFT particles, is mutated in a consanguineous family with Bardet-Biedl syndrome.
IFT27, encoding a small GTPase component of IFT particles, is mutated in a consanguineous family with Bardet-Biedl syndrome.
Human molecular genetics (2014-02-04)
Mohammed A Aldahmesh, Yuanyuan Li, Amal Alhashem, Shams Anazi, Hisham Alkuraya, Mais Hashem, Ali A Awaji, Sameera Sogaty, Abdullah Alkharashi, Saeed Alzahrani, Selwa A Al Hazzaa, Yong Xiong, Shanshan Kong, Zhaoxia Sun, Fowzan S Alkuraya
ABSTRACT
Bardet-Biedl syndrome (BBS) is an autosomal recessive ciliopathy with multisystem involvement. So far, 18 BBS genes have been identified and the majority of them are essential for the function of BBSome, a protein complex involved in transporting membrane proteins into and from cilia. Yet defects in the identified genes cannot account for all the BBS cases. The genetic heterogeneity of this disease poses significant challenge to the identification of additional BBS genes. In this study, we coupled human genetics with functional validation in zebrafish and identified IFT27 as a novel BBS gene (BBS19). This is the first time an intraflagellar transport (IFT) gene is implicated in the pathogenesis of BBS, highlighting the genetic complexity of this disease.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Pricing and availability is not currently available.