MilliporeSigma
  • Regional mapping of the human gene encoding the novel pituitary polypeptide 7B2 to chromosome 15q13----q14 by in situ hybridization.
Regional mapping of the human gene encoding the novel pituitary polypeptide 7B2 to chromosome 15q13----q14 by in situ hybridization.
Cytogenetics and cell genetics (1989-01-01)
A J Roebroek, M R Dehaen, A van Bokhoven, G J Martens, P Marÿnen, H van den Berghe, W J Van de Ven
ABSTRACT
Genetic sequences encoding the novel pituitary polypeptide 7B2 were isolated from a human pituitary cDNA library. Hybridization analysis of a panel of human x mouse cell hybrids with a 7B2 cDNA probe indicated that the locus for the human 7B2 gene is probably located on chromosome 15. In situ hybridization analysis of metaphase chromosomes allowed the regional localization of the 7B2 gene to chromosome 15 at q13----q14.