MilliporeSigma
Characterization of the human analogue of a Scrapie-responsive gene.
The Journal of biological chemistry (1998-07-11)
M Dron, F Dandoy-Dron, F Guillo, L Benboudjema, J J Hauw, P Lebon, D Dormont, M G Tovey
ABSTRACT
We have recently described a novel mRNA denominated ScRG-1, the level of which is increased in the brains of Scrapie-infected mice (Dandoy-Dron, F., Guillo, F., Benboudjema, L., Deslys, J.-P., Lasmézas, C., Dormont, D., Tovey, M. G., and Dron, M. (1998) J. Biol. Chem. 273, 7691-7697). The increase in ScRG-1 mRNA in the brain follows the accumulation of PrPSc, the proteinase K-resistant form of the prion protein (PrP), and precedes the widespread neuronal death that occurs in late stage disease. In the present study, we have isolated a cDNA encoding the human counterpart of ScRG-1. Comparison of the human and mouse transcripts firmly established that both sequences encode a highly conserved protein of 98 amino acids that contains a signal peptide, suggesting that the protein may be secreted. Examination of the distribution of human ScRG-1 mRNA in adult and fetal tissues revealed that the gene was expressed primarily in the central nervous system as a 0.7-kilobase message and was under strict developmental control.