MilliporeSigma
  • Somatic cell mapping of the human cyclophilin B gene (PPIB) to chromosome 15.
Somatic cell mapping of the human cyclophilin B gene (PPIB) to chromosome 15.
Cytogenetics and cell genetics (1992-01-01)
L B Peddada, J D McPherson, R Law, J J Wasmuth, P Youderian, R J Deans
ABSTRACT
The polymerase chain reaction (PCR) technique was used to generate a unique probe complementary to the hydrophobic 5' end of the human cyclophilin B gene. This unique probe was hybridized to DNAs from human x hamster hybrid somatic cell lines retaining different combinations of human chromosomes. The gene was assigned to chromosome 15.