MilliporeSigma
  • Ubiquinol-10 supplementation activates mitochondria functions to decelerate senescence in senescence-accelerated mice.
Ubiquinol-10 supplementation activates mitochondria functions to decelerate senescence in senescence-accelerated mice.
Antioxidants & redox signaling (2013-10-16)
Geng Tian, Jinko Sawashita, Hiroshi Kubo, Shin-ya Nishio, Shigenari Hashimoto, Nobuyoshi Suzuki, Hidekane Yoshimura, Mineko Tsuruoka, Yaoyong Wang, Yingye Liu, Hongming Luo, Zhe Xu, Masayuki Mori, Mitsuaki Kitano, Kazunori Hosoe, Toshio Takeda, Shin-ichi Usami, Keiichi Higuchi
ABSTRACT
The present study was conducted to define the relationship between the anti-aging effect of ubiquinol-10 supplementation and mitochondrial activation in senescence-accelerated mouse prone 1 (SAMP1) mice. Here, we report that dietary supplementation with ubiquinol-10 prevents age-related decreases in the expression of sirtuin gene family members, which results in the activation of peroxisome proliferator-activated receptor γ coactivator 1α (PGC-1α), a major factor that controls mitochondrial biogenesis and respiration, as well as superoxide dismutase 2 (SOD2) and isocitrate dehydrogenase 2 (IDH2), which are major mitochondrial antioxidant enzymes. Ubiquinol-10 supplementation can also increase mitochondrial complex I activity and decrease levels of oxidative stress markers, including protein carbonyls, apurinic/apyrimidinic sites, malondialdehydes, and increase the reduced glutathione/oxidized glutathione ratio. Furthermore, ubiquinol-10 may activate Sirt1 and PGC-1α by increasing cyclic adenosine monophosphate (cAMP) levels that, in turn, activate cAMP response element-binding protein (CREB) and AMP-activated protein kinase (AMPK). These results show that ubiquinol-10 may enhance mitochondrial activity by increasing levels of SIRT1, PGC-1α, and SIRT3 that slow the rate of age-related hearing loss and protect against the progression of aging and symptoms of age-related diseases.
Sigma-Aldrich
Quantification kit for oxidized and reduced glutathione, 200 tests
Pricing and availability is not currently available.