MilliporeSigma
  • Modulation of redox signaling promotes apoptosis in epithelial ovarian cancer cells.
Modulation of redox signaling promotes apoptosis in epithelial ovarian cancer cells.
Gynecologic oncology (2011-05-31)
Zhongliang Jiang, Nicole M Fletcher, Rouba Ali-Fehmi, Michael P Diamond, Husam M Abu-Soud, Adnan R Munkarah, Ghassan M Saed
ABSTRACT
Epithelial ovarian cancer (EOC) cells are known to be resistant to apoptosis through a mechanism that may involve alteration in their redox balance. NADPH oxidase is a major source of intracellular superoxide, which is converted to the less toxic product by superoxide dismutase (SOD). Superoxide contributes to hypoxia inducible factor (HIF)-1α stabilization. We sought to determine the effects of inhibiting the generation of intracellular reactive oxygen species (ROS) on apoptosis of EOC cells. Diphenyleneiodonium (DPI), an irreversible ROS inhibitor, was used to inhibit the generation of ROS in EOC cell lines, SKOV-3 and MDAH-2774, followed by assessment of apoptosis, NADPH oxidase, SOD3 and HIF-1α expression. A combination of immunohistochemistry, immunoprecipitation/western blot, and real-time RT-PCR were utilized to evaluate the expression of these enzymes in EOC cells as well as normal ovarian tissue and ovarian cancer tissue specimens. DPI treatment significantly induced apoptosis in both EOC cell lines as evident by increased caspase-3 activity and TUNEL assay. Additionally, both EOC cell lines were found to express NADPH oxidase, HIF-1α, and SOD3, which were highly sensitive to DPI treatment. DPI treatment resulted in reduced NADPH oxidase, SOD3 and HIF-1α levels. Furthermore, ovarian cancer tissues were found to manifest higher NADPH oxidase levels as compared to normal ovarian tissues. These data suggest that lowering oxidative stress, possibly through the inhibition of NADPH oxidase, induces apoptosis in ovarian cancer cells and may serve as a potential target for cancer therapy.