MilliporeSigma
  • Effective reversal of edoxaban-associated bleeding with four-factor prothrombin complex concentrate in a rabbit model of acute hemorrhage.
Effective reversal of edoxaban-associated bleeding with four-factor prothrombin complex concentrate in a rabbit model of acute hemorrhage.
Anesthesiology (2014-11-25)
Eva Herzog, Franz Kaspereit, Wilfried Krege, Baerbel Doerr, Jochen Mueller-Cohrs, Ingo Pragst, Yoshiyuki Morishima, Gerhard Dickneite
ABSTRACT
Edoxaban is an oral, selective direct factor Xa inhibitor approved in Japan for venous thromboembolism prevention after orthopedic surgery. Data are lacking regarding reversal strategies for edoxaban; this study assessed whether four-factor prothrombin complex concentrate (Beriplex/Kcentra; CSL Behring GmbH, Marburg, Germany) can effectively reverse its effects on hemostasis using a previously described rabbit model. The study comprised assessments of thrombin generation in vitro, pharmacokinetic parameters, and edoxaban reversal in vivo. In a blinded in vivo stage, a standardized kidney incision was performed in animals (n = 11 per group) randomized to receive vehicle + saline, edoxaban (1,200 μg/kg) + saline, or edoxaban (1,200 μg/kg) + four-factor prothrombin complex concentrate (50 IU/kg). Animals were monitored for treatment impact on hemostasis and coagulation parameters. Data are median (range). Statistical tests were adjusted for multiple testing. Edoxaban administration increased blood loss (30 [2 to 44] ml) and time to hemostasis (23 [8.5 to 30.0] min) compared with the control group (3 [1 to 8] ml and 3 [2.0 to 5.0] min, respectively). Biomarkers of coagulation (prothrombin time, activated partial thromboplastin time, whole blood clotting time) and thrombin generation parameters (e.g., peak thrombin, endogenous thrombin potential, lag time) were also affected by edoxaban. Administration of four-factor prothrombin complex concentrate significantly reduced time to hemostasis (to 8 [6.5 to 14.0] min, observed P < 0.0001) and total blood loss (to 9 [4 to 22] ml, observed P = 0.0050) compared with the edoxaban + saline group. Of the biomarkers tested, prothrombin time, whole blood clotting time, and endogenous thrombin potential correlated best with clinical parameters. In a rabbit model of hemostasis, four-factor prothrombin complex concentrate administration significantly decreased edoxaban-associated hemorrhage.
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Water, for TOC analysis
Sigma-Aldrich
D-(+)-Glucose, SAJ special grade, ≥98.0%
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus