MilliporeSigma
  • Prucalopride is no more effective than placebo for children with functional constipation.
Prucalopride is no more effective than placebo for children with functional constipation.
Gastroenterology (2014-09-23)
Suzanne M Mugie, Bartosz Korczowski, Piroska Bodi, Alexandra Green, René Kerstens, Jannie Ausma, Magnus Ruth, Amy Levine, Marc A Benninga
ABSTRACT
Prucalopride is a selective, high-affinity agonist of the 5-hydroxytryptamine (serotonin) receptor 4 that enhances motility in the gastrointestinal tract. We performed a multicenter, randomized, placebo-controlled, double-blind, phase 3 trial to evaluate the efficacy and safety of prucalopride in children (6 months to 18 years old) with functional constipation. Children with functional constipation, based on the Rome III criteria, were given prucalopride (children ≤ 50 kg were given a 0.04 mg/kg oral solution; children >50 kg were given a 2-mg tablet) or placebo once daily for 8 weeks. The primary efficacy end point was the proportion of children with toileting skills who had a mean of ≥ 3 spontaneous bowel movements/week and ≤ 1 episode of fecal incontinence/2 weeks, from study weeks 5-8 (responders). Adverse events, clinical laboratory values, and electrocardiograms were monitored. Efficacy and safety were assessed in 213 children (106 prucalopride, 107 placebo). Twenty-five percent were younger than 4 years old, 50% were 4-11 years old, and 25% were 12-18 years old; 55.4% were girls. At screening, 62.3% of patients in the prucalopride group and 55.1% in the placebo group had a history of fecal incontinence; 60.4% and 55.1% in the prucalopride and placebo groups, respectively, had a mean of ≤ 1 spontaneous bowel movements/week. The proportion of responders was similar between groups (prucalopride, 17.0% and placebo, 17.8%). There were no statistically significant differences in the primary efficacy end point when patients were stratified by sex, age group, or country. The incidence of treatment-emergent adverse events was similar in the prucalopride (69.8%) and placebo (60.7%) groups. Prucalopride, although generally well tolerated, was not more effective than placebo in children with functional constipation. ClinicalTrials.gov Number: NCT01330381.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, FCC, FG
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard