MilliporeSigma
  • Active vitamin D possesses beneficial effects on the interaction between muscle and bone.
Active vitamin D possesses beneficial effects on the interaction between muscle and bone.
Biochemical and biophysical research communications (2014-06-14)
Ken-ichiro Tanaka, Ippei Kanazawa, Toru Yamaguchi, Shozo Yano, Hiroshi Kaji, Toshitsugu Sugimoto
ABSTRACT
Vitamin D deficiency and advanced glycation end products (AGEs) are suggested to be involved in the pathogenesis of osteoporosis and sarcopenia. However, the effects of vitamin D and AGEs on myogenesis and the interaction between muscle and bone remains still unclear. We previously showed that osteoglycin (OGN) is secreted from myoblasts and stimulates osteoblastic differentiation, suggesting that it plays important roles in the interaction between muscle and bone. The aim of this study is thus to examine the effects of vitamin D and AGEs on myoblastic differentiation of C2C12 cells and osteoblastic differentiation of osteoblastic MC3T3-E1 cells through OGN expression. 1α,25-dihydroxyvitamin D3 (1,25D) and eldecalcitol, an active vitamin D analog, induced the expression of MyoD, myogenin and OGN, and these effects were abolished by vitamin D receptor (VDR) suppression by siRNA in C2C12 cells. Moreover, conditioned medium from 1,25D-pretreated C2C12 cells stimulated the expression of type 1 collagen and alkaline phosphatase in MC3T3-E1 cells, compared to control medium from 1,25D-untreated C2C12 cells. In contrast, conditioned medium from VDR-suppressed and 1,25D-pretreated C2C12 cells showed no effects. AGE2 and AGE3 suppressed the expression of MyoD, myogenin and OGN in C2C12 cells. Moreover, 1,25D blunted the AGEs' effects. In conclusion, these findings showed for the first time that active vitamin D plays important roles in myogenesis and muscle-induced osteoblastogenesis through OGN expression. Active vitamin D treatment may rescue the AGEs-induced sarcopenia as well as - suppressed osteoblastic differentiation via OGN expression in myoblasts.
Sigma-Aldrich
MISSION® esiRNA, targeting human CYP27B1
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human VDR
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Pricing and availability is not currently available.