MilliporeSigma
  • Improved stability of TMS derivatives for the robust quantification of plant polar metabolites by gas chromatography-mass spectrometry.
Improved stability of TMS derivatives for the robust quantification of plant polar metabolites by gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-23)
Anthony Quéro, Cyril Jousse, Michelle Lequart-Pillon, Eric Gontier, Xavier Guillot, Bernard Courtois, Josiane Courtois, Corinne Pau-Roblot
ABSTRACT
Plant metabolite profiling is commonly carried out by GC-MS of methoximated trimethylsilyl (TMS) derivatives. This technique is robust and enables a library search for spectra produced by electron ionization. However, recent articles have described problems associated with the low stability of some TMS derivatives. This limits the use of GC-MS for metabolomic studies that need large sets of qualitative and quantitative analyses. The aim of this work is to determine the experimental conditions in which the stability of TMS derivatives could be improved. This would facilitate the analysis of the large-scale experimental designs needed in the metabolomics approach. For good repeatability, the sampling conditions and the storage temperature of samples during analysis were investigated. Multiple injections of one sample from one vial led to high variations while injection of one sample from different vials improved the analysis. However, before injection, some amino acid TMS derivatives were degraded during the storage of vials in the autosampler. Only 10% of the initial quantity of glutamine 3 TMS and glutamate 3 TMS and 66% of α-alanine 2 TMS was detected 48 h after derivatization. When stored at 4 °C until injection, all TMS derivatives remained stable for 12 h; at -20 °C, they remained stable for 72 h. From the integration of all these results, a detailed analytical procedure is thus proposed. It enables a robust quantification of polar metabolites, useful for further plant metabolomics studies using GC-MS.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%